Learn More
GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets
Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. GE Healthcare Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Brand:  GE Healthcare 10600078
Additional Details : Weight : 0.07700kg
 View more versions of this product
Product Code. 15239854
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
Individual sheets.
  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
Specifications
Specifications
25/Pk.
10cm
Sheets
10cm
Western blots, dot-blot assays, and other protein or nucleic acid methods
0.45μm
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Amersham Protran Premium
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement