Learn More
GE Healthcare Dharmacon siGLO™ Red Transfection Indicator
Chemically modified to localize to the nucleus and significantly extend fluorescence as a qualitative indicator of delivery and should not be used for quantitative determination of siRNA uptake.
Brand:  GE Healthcare Dharmacon D-001630-02-50
Additional Details : Weight : 0.00500kg
 View more versions of this product
Product Code. 15509792
  • 8454.78 SEK / 50nmol
Estimated Shipment date
from Supplier 20-02-2018
Add to basket
Specifications
Specifications
Cy3 Thodamine or Polyethylene
Determination of optimal siRNA transfection conditions in mammalian cells
50nm
Videos