Learn More
Cytiva GST Bulk Kit
For small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins
Brand: Cytiva 27-4570-01
Additional Details : Weight : 0.50000kg
Description
- Gentle elution conditions preserve protein antigenicity and function
- Flexible GST-glutathione affinity system for rapid column or batch purification
- High specificity (compared to histidine-tagged purification)
- Provide reagents for induction expression and elution of fusion proteins
- Sufficient reagents to purify up to 50mg fusion protein
- Includes 10mL bulk pack of Glutathione Sepharose™ 4B, 5 disposible columns, GST buffer kit and 500 mg Isopropyl Β-D Thiogalactoside
Specifications
GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged protein | |
20°C to 25°C | |
Chromatography Media |
10 mL bulk pack of Glutathione Sepharose 4B medium, 5 x gravity-flow columns, IPTG, GST Buffer Kit | |
1 Kit |