Learn More
Cytiva (Formerly GE Healthcare Life Sciences) illustra™ ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
Brand:  Cytiva (Formerly GE Healthcare Life Sciences) US79010
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15209764
  • 638.58 SEK / 100 reactions
Estimated Shipment
04-11-2020
Add to basket
Description
Description
  • Easy to automate
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Complete heat inactivation of the enzymes within 10 minutes
  • Fast 15 minute protocol
  • Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
  • Improves digestion efficiency with no degradation of the target PCR product
  • Sample is completely intact and ready for immediate use in manual or automated processes
Specifications
Specifications
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.
illustra ExoProStar S
100 Reactions