missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) NHS Mag Sepharose™

NHS Mag Sepharose magnetic beads for convenient enrichment of selected proteins using pull-down sample preparation techniques

Brand:  Cytiva (Formerly GE Healthcare Life Sciences) 28-9513-80

Additional Details : Weight : 0.04000kg Transport : UN number : 1219 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 11520894

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity. An antibody or other protein with desired affinity is covalently coupled to the medium via primary amines for the purpose of capture of target proteins of interest.

  • Visible and dense Sepharose based magnetic beads with N-hydroxysuccinimide (NHS) ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/pull-down of target proteins - only low amounts of bound capture molecule
  • Flexible protocols with elution conditions optimized for downstream applications such as electrophoresis and MS

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: N-hydroxysuccinimide
Ligand density: 8-14μmol/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% 2-propanol
Storage temperature: 4°C to 8°C

Specifications

Specifications

NHS Mag Sepharose
Designed for covalent coupling of any Protein or biomolecule containing a free Amine group and is used for various types of pull down experiments, for biomarker research
8 to 14μM/mL
37 to 100μm
4° to 8°C
8 to 14μM/mL
N-hydroxysuccinimide
Paramagnetic spherical, highly cross linked agarose particles
4 x 500μL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NHS Mag Sepharose, 4 x 500 ul

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • UNKNOWN AQUATIC-Contains 15% of components with unknown hazards to the aquatic environment.

Documents