missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare PhastGel™ Precast Gels

Select from homogeneous and gradient polyacrylamide gels. GE Healthcare PhastGel™ Precast Gels save time and improve reproducibility.

1627.00 SEK - 3200.00 SEK

Specifications

Width (Metric) Gel 5 cm
For Use With (Application) Used for industrial applications such as analytical chemistry and research
Storage Requirements 4 - 8°C
Gel Thickness 0.45 mm
Material GelBond polyester film
View More Specs

Products 9
Product Code Brand Formulation Height (Metric) Gel Percentage Separation Range Price Quantity & Availability  
Product Code Brand Formulation Height (Metric) Gel Percentage Separation Range Price Quantity & Availability  
11738458 View Documents GE Healthcare
17-0540-01
4.3 cm 10 to 15% 80 to 400 Mr 1722.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
10309294 View Documents GE Healthcare
17-0542-01
4.4 cm 8 to 25% 50 to 30 Mr 1722.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
11798458 View Documents GE Healthcare
17-0622-01
4.9 cm 7.5% 70 to 230 Mr 2102.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
11718468 View Documents GE Healthcare
17-0624-01
5.1 cm 20% 10 to 50 Mr 2102.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
10734927 View Documents GE Healthcare
17-0678-01
4.5 cm 4 to 15% 70 Mr 2102.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
11798468 View Documents GE Healthcare
17-0679-01
High-density, 20% Polyacrylamide, 30% Ethylene Glycol 4.3 cm 7.5% 20 to 60 Mr 3200.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
11708468 View Documents GE Healthcare
17-0623-01
Gel with 12.5% Polyacrylamide 5 cm 12.5% 230 Mr 2102.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
10678225 View Documents GE Healthcare
17-0543-01
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.6 cm 5% T, 3% C 3 to 9 Mr 1627.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
10-02-2020
Add to basket
 
10574375 View Documents GE Healthcare
17-0677-01
Containing 2 to 6% Pharmalyte as carrier Ampholyte 4.3 cm 5% T, 3% C 10 to 50 Mr 2102.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
Description

Description

  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing and native
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling

PhastGel homogenous gels

  • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
  • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution

PhastGel homogenous gels

  • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
Specifications

Specifications

5 cm
4 - 8°C
GelBond polyester film
Used for industrial applications such as analytical chemistry and research
0.45 mm