missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare PlusOne™ Silver Staining Kit for Protein

Use for reproducible and sensitive staining of proteins in polyacrylamide gels. PlusOne™ Silver Staining Kit for Protein enables staining of proteins in less than two hours.

Brand:  GE Healthcare 17-1150-01

Additional Details : Weight : 0.92120kg Transport : UN number : 2209 Chem class : 8 Pack group : III


Product Code. 10105944

  • 1898.80 SEK / 634g


Estimated Shipment
10-02-2020
Add to basket


Includes: Sodium acetate, 5% sodium thiosulfate, EDTA-Na2, sodium carbonate, 2.5% silver nitrate, 37% formaldehyde, and 25% glutaraldehyde

Description

Description

  • Typical sensitivity: 0.2 to 0.6ng protein/band
  • Use with 2-D electrophoresis and mass spectrometry by omitting glutaraldehyde from staining procedure

Order Info

Components supplied in quantities sufficient to stain 10 precast 12.5 × 26cm gels, 20 minigels (8-10 × 8cm), or 30 to 60 PhastGel™ Gels.

Specifications

Specifications

1 Kit
For use in Hoefer automated gel stainer as well as for manual silver staining. Silver staining is a highly sensitive method for visualizing proteins separated in SDS-polyacrylamide gels.
10 to 30°C
Protein Silver Staining Kit
PlusOne Silver Staining Kit, Protein and PlusOne DNA Silver Staining Kit are two silver staining kits that bring speed, convenience, and very high sensitivity to the staining of proteins and nucleic acids in polyacrylamide electrophoresis. gels.
Sodium Acetate: 10 x 17g packets, 5% Sodium Thiosulfate: 1 x 100mL bottle, EDTA-Na2: 10 x 3.65g packets, Sodium Carbonate: 10 x 6.25g packets, 2.5% Silver Nitrate: 2 x 125mL bottles, 37% Formaldehyde: 1 x 2.5mL bottle, 25% Glutaraldehyde: 1 x 12.5mL bottle
10 DALT precast gels, 10 precast 12.5 x 26cm gels, 20 mini slab gels (8 to 10 x 8cm) or 30 to 60 PhastGel™
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Respiratory sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H400-Very toxic to aquatic life.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations..
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305-IF IN EYES:Immediately call a POISON CENTER or physician.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : Glycine, N, N'-1, 2-ethanediylbis¢N-(carboxymethyl)-, sodiumsalt, hydrate (1:2:2), formaldehyde, gl

Documents