Läs mer
GE Healthcare TE 22 Mini Tank Transfer Unit
Performs protein and nucleic acid transfers for Western, Southern, and Northern blots in less than one hour
Brand:  GE Healthcare 80620426
Produktkod. 12971630
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Innehåller: Four gel cassettes; eight foam sponges, 3mm thick; four foam sponges, 6mm thick; and 25 sheets of blotter paper
Beskrivning
Beskrivning
  • Transfers four mini-gels up to 9 × 10cm in size in about 1L of buffer
  • Built-in alumina-covered heat exchanger dissipates heat generated by tank electrotransfer; typical temperature increase: ≤5°C
  • 80cm of wire in seven parallel strands on each electrode panel delivers a uniform electrical field
  • Hinged, color-coded cassettes allow quick setup of transfers
  • Replacement electrode panel, sponges, and gel cassettes available separately
TRUSTED_SUSTAINABILITY
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer