Läs mer
Gedunin, TRC
Produktkod. 30138406
Change view
Kvantitet:
1 mg
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138406 25 mg -
30135880 2.5 mg 2.5mg
30009175 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138406 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG30350325MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2753-30-2
Molekylformel C28 H34 O7
Molekylvikt (g/mol) 482.56
Synonym Gedunin, (1S,3aS,4aR,4bS,5R,6aR,10aR,10bR,12aS)-5-(Acetyloxy)-1-(3-furanyl)-1,5,6,6a,7,10a,10b,11,12,12a-decahydro-4b,7,7,10a,12a-pentamethyloxireno[c]phenanthro[1,2-d]pyran-3,8(3aH,4bH)-dione, Gedunine, NSC 113497, (5α,7α,13α,14β,15β,17aα)-7-(Acetyloxy)-14,15:21,23-diepoxy-4,4,8-trimethyl-D-homo-24-nor-17-oxachola-1,20,22-triene-3,16-dione
LEDER CC(=O)O[C@@H]1C[C@H]2C(C)(C)C(=O)C=C[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](OC(=O)[C@H]5O[C@@]45[C@]13C)c6cocc6
Specifikationer
Kemiskt namn eller material Gedunin
Kvantitet 25 mg
InChI formel InChI=1S/C28H34O7/c1-15(29)33-20-13-18-24(2,3)19(30)8-10-25(18,4)17-7-11-26(5)21(16-9-12-32-14-16)34-23(31)22-28(26,35-22)27(17,20)6/h8-10,12,14,17-18,20-22H,7,11,13H2,1-6H3/t17-,18+,20-,21+,22-,25-,26+,27+,28-/m1/s1
Formel vikt 482.2305