Läs mer
Fisherbrand™ Gel Casting Tray
Produktkod. p-8007108
Change view
Längd (metrisk) platta:
75 mm
115 mm
130 mm
200 mm
300 mm
Bredd (metrisk):
60 mm
100 mm
128 mm
200 mm
250 mm
Förpackningsstorlek:
Varje
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Längd (metrisk) platta Bredd (metrisk) unitSize
10142041 200 mm 200 mm Varje
10569591 130 mm 128 mm Varje
11709665 300 mm 250 mm Varje
10395541 75 mm 60 mm Varje
10479352 115 mm 100 mm Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 10142041 Leverantör Fisherbrand™ Leverantörsnummer HU20UT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Forms leak proof seal against inner walls.
  • Silicone gaskets to line casting tray
  • Forms leak proof seal against inner walls of running chamber when casting tray is turned at 90° to direction of electrophoresis
Specifikationer
Typ Gel Casting Tray
Inkluderar Leakproof Silicone Seals
Material Acrylic
Längd (metrisk) platta 200 mm
Bredd (metrisk) 200 mm