Läs mer
Fisherbrand™ Gel Casting Trays for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System
Produktkod. 11776155
Längd (metrisk) platta:
70 mm
100 mm
150 mm
Förpackningsstorlek:
Varje
Produktkod. 11776155
Brand: Fisherbrand™ TVGGT7
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Accessory for use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System.
  • UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
  • Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
  • aluminum casting gates - ensure easy casting of the gel inside or outside of the tank.They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
  • Available in three different dimensions
Specifikationer
Typ Gel Casting Tray
Längd (metrisk) platta 70 mm
Bredd (metrisk) 100 mm