Läs mer
Fisherbrand™ Gel Casting Trays for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System
Produktkod. 11786155
Change view
Längd (metrisk) platta:
70 mm
100 mm
150 mm
Förpackningsstorlek:
Varje
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Längd (metrisk) platta unitSize
11786155 100 mm Varje
11796155 150 mm Varje
11776155 70 mm Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11786155 Leverantör Fisherbrand™ Leverantörsnummer TVGGT10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Accessory for use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System.
  • UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
  • Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
  • aluminum casting gates - ensure easy casting of the gel inside or outside of the tank.They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
  • Available in three different dimensions
Specifikationer
Typ Gel Casting Tray
Längd (metrisk) platta 100 mm
Bredd (metrisk) 100 mm