Läs mer
Biodesign™ Gel storage system GelFile binder and 20 gel protector sheets 4 gels per sheet for gels up to 115mm x 180mm BioDesign
Beskrivning
Stores, organises and displays dried polyacrylamide and agarose gels
- Each BioDesign GelFile™ binder contains heavy crystal clear vinyl pages with pockets and white backgrounds
- The pages are chemically neutral and are guaranteed not to alter your dried gels or turn yellow for TEN years
- Dried gels are held flat for long term storage without curling
- Shields gels from moisture, accidental spills and possible damage
- White background allows optimal comparison between gels
- Each BioDesign GelFile™ binder holds 20 to 25 pages
- A clear label holder on the spine allows you to personalise each binder
Specifikationer
Specifikationer
| Gel storlek | 11,5 x 18 cm |
| Kvantitet | 1 Binder, 20 Gel Protector Sheets |
| Produkttyp | Gel Storage System, Gel Storage Binder |