Learn More
Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix
Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix
425.00 SEK - 1904.00 SEK
Specifications
Format Tube(s)
Product Type Basement Membrane Matrix
Sterility Sterile
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11692139
View Documents
Gibco™
A1413201
1mL
425.00 SEK
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 06-10-2021
12053569
View Documents
Gibco™
A1413202
5mL
1904.00 SEK
5mL
View alternative products
Save Up to 
»
Estimated Shipment: 26-10-2021
Description
Description
Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors and is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation. The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans. It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC). Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors. As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format.
Key Features of Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix:
  • Free of lactose dehydrogenase elevating virus (LDEV) making it ideal for all types of cell culture and mouse in vivo applications
  • Consistent protein concentration lot-to-lot helps reduce the need to screen lots
  • Protocols for various applications, including tube formation assays, are available
    Gain Reliability and Consistency in Your hESC Studies
    Recent advances in processing EHS tumor cells have resulted in virus free basement membrane extracts. Every lot is tested by RT-PCR methods so you can now have confidence that every lot of Geltrex™ is virus free. Additionally, our consistent manufacturing process helps ensure a protein concentration with very little variation from lot-to-lot, averaging 15 mg/ml so you can have confidence in every purchase.
    Investigate Angiogenesis with Validated Gibco™ Protocols
    There are many applications for Geltrex™ LDEV-Free reduced growth factor basement membrane matrix, each requiring different thicknesses and concentrations. In general, a protein concentration of >5 mg/ml is used for differentiation studies of primary cells. Geltrex™ LDEV-Free reduced growth factor basement membrane matrix has been used to develop tube formation assay protocols using Gibco™ primary cells and specialty media. See this and other angiogenesis assay protocols
  • Specifications
    Specifications
    Tube(s)
    Basement Membrane Matrix
    Sterile
    SDS
    Product Certifications
    For Research Use Only. Not for use in diagnostic procedures.