missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Gemcabene, MedChemExpress
Description
Gemcabene (PD-72953), a first-in-class lipid-lowering agent, lowers low-density lipoprotein cholesterol (LDL-C), decreases triglycerides, and raises high-density lipoprotein cholesterol (HDL-C) and lowers pro-inflammatory acute-phase protein, C-reactive protein (CRP), exerting anti-inflammatory activity.
Specifications
Specifications
| Kemiskt namn eller material | Gemcabene |
| Fysisk form | Solid |
| CAS | 183293-82-5 |
| Färg | White |
| Kvantitet | 10 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C16H30O5 |
| För användning med (applikation) | Neuroscience-Neuromodulation |
| Synonym | PD-72953 |
| Löslighetsinformation | DMSO : 100 mg/mL (330.68 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?