Läs mer
Gemcitabine-13C,15N2 Hydrochloride, TRC
Produktkod. 18053630
Change view
Kvantitet:
0.5 mg
1 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18053630 1 mg
18030920 10 mg
18022530 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18053630 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG3050021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1262897-74-4
Molekylformel C813CH12ClF2N15N2O4
Molekylvikt (g/mol) 302.64
Synonym 2’-Deoxy-2’,2’-difluorocytidine-13C,15N2 Hydrochloride;
LEDER FC1(F)[C@H](O)[C@@H](CO)O[C@H]1[15N]2[13C]([15N]=C(N)C=C2)=O.Cl
Specifikationer
Kemiskt namn eller material Gemcitabine-13C,15N2 Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C9H11F2N3O4.ClH/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17;/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17);1H/t4-,6-,7+;/m1./s1/i8+1,13+1,14+1;
Procent renhet >95%