Läs mer
Gemcitabine Diphosphate Choline, TRC
Produktkod. 30407998
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30407998 1 mg
1 options
Produktkod. 30407998 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG3050151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Gemcitabine Diphosphate Choline
CAS 1643126-46-8
Molekylformel C14 H24 F2 N4 O10 P2
Kvantitet 1 mg
Synonym Cytidine 5'-(trihydrogen diphosphate), 2'-deoxy-2',2'-difluoro-, P'-[2-(trimethylammonio)ethyl] ester, inner salt
InChI formel InChI=1S/C14H24F2N4O10P2/c1-20(2,3)6-7-27-31(23,24)30-32(25,26)28-8-9-11(21)14(15,16)12(29-9)19-5-4-10(17)18-13(19)22/h4-5,9,11-12,21H,6-8H2,1-3H3,(H3-,17,18,22,23,24,25,26)/t9-,11-,12-/m1/s1
LEDER C[N+](C)(C)CCOP(=O)([O-])OP(=O)(O)OC[C@H]1O[C@@H](N2C=CC(=NC2=O)N)C(F)(F)[C@@H]1O
IUPAC-namn [[(2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-4,4-difluoro-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
Molekylvikt (g/mol) 508.31
Formel vikt 508.0936