Learn More
Applied Biosystems™ GeneScan™ 350 ROX™ dye Size Standard
GeneScan™350 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence based DNA electrophoresis systems.
Brand: Applied Biosystems™ 401735
Additional Details : Weight : 0.02950kg
Includes: Contains 8 ROX labeled single stranded DNA fragments for use as a standard
Description
- The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs
Order Info
Each
Specifications
Contains 8 ROX labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze). | |
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer | |
ROX | |
GeneScan™, ROX™ | |
Wet Ice |
Standard | |
Sequencing | |
800 Reactions | |
Dye | |
Standard |
For research use only