Learn More
Applied Biosystems™ GeneScan™ 350 ROX™ dye Size Standard
GeneScan™350 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence based DNA electrophoresis systems.
Brand:  Applied Biosystems™ 401735
Additional Details : Weight : 0.02950kg
Product Code. 16385242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Contains 8 ROX labeled single stranded DNA fragments for use as a standard
Description
Description
  • The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs
Order Info
Each
Specifications
Specifications
Contains 8 ROX labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze).
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
ROX
GeneScan™, ROX™
Wet Ice
Standard
Sequencing
800 Reactions
Dye
Standard
Special Offers
For research use only