missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin sulfate solution-10, MP Biomedicals™

Catalog Number 11411832
Click to view available options
Quantity:
10 mL
100 mL
5 x 10 mL
This item is not returnable. View return policy
1405-41-0
MFCD00270181
Garamycin, Genoptic
This item is not returnable. View return policy
1405-41-0
MFCD00270181
Garamycin, Genoptic
Type Gentamicin Sulfate
Physical Form Liquid
Grade USP
Color Colorless to Yellow
Quantity 100 mL
Chemical Name or Material Gentamycin Sulfate Solution - 10, Cell Biology and Analysis
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'