Läs mer
Gepirone, TRC
Produktkod. 30405616
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405616 10 mg
30399087 100 mg
30391845 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30405616 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG36500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83928-76-1
Molekylformel C19 H29 N5 O2
Molekylvikt (g/mol) 359.47
Synonym 2,6-Piperidinedione, 4,4-dimethyl-1-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, 4,4-Dimethyl-1-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-2,6-piperidinedione, Gepirone
IUPAC-namn 4,4-dimethyl-1-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]piperidine-2,6-dione
LEDER CC1(C)CC(=O)N(CCCCN2CCN(CC2)c3ncccn3)C(=O)C1
Specifikationer
Kemiskt namn eller material Gepirone
Kvantitet 10 mg
InChI formel InChI=1S/C19H29N5O2/c1-19(2)14-16(25)24(17(26)15-19)9-4-3-8-22-10-12-23(13-11-22)18-20-6-5-7-21-18/h5-7H,3-4,8-15H2,1-2H3
Formel vikt 359.2321