Learn More
Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%, Thermo Scientific Chemicals
Catalog Number p-7024467
Click to view available options
Quantity:
25 g
100 g
689-67-8
C13H22O
194.318
MFCD00008910
HNZUNIKWNYHEJJ-FMIVXFBMSA-N
geranylacetone, geranyl acetone, e-6,10-dimethylundeca-5,9-dien-2-one, trans-geranylacetone, 5e-6,10-dimethylundeca-5,9-dien-2-one, geranylacetone, trans, dihydropseudoionone, 6,10-dimethylundeca-5,9-dien-2-one, e-6,10-dimethyl-5,9-undecadien-2-one, unii-9b7ry79u9z
1549778
CHEBI:67206
(5E)-6,10-dimethylundeca-5,9-dien-2-one
CC(=CCCC(=CCCC(=O)C)C)C
689-67-8
C13H22O
194.318
MFCD00008910
HNZUNIKWNYHEJJ-FMIVXFBMSA-N
geranylacetone, geranyl acetone, e-6,10-dimethylundeca-5,9-dien-2-one, trans-geranylacetone, 5e-6,10-dimethylundeca-5,9-dien-2-one, geranylacetone, trans, dihydropseudoionone, 6,10-dimethylundeca-5,9-dien-2-one, e-6,10-dimethyl-5,9-undecadien-2-one, unii-9b7ry79u9z
1549778
CHEBI:67206
(5E)-6,10-dimethylundeca-5,9-dien-2-one
CC(=CCCC(=CCCC(=O)C)C)C
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refregirated.
TRUSTED_SUSTAINABILITY
Density 0.87
Boiling Point 250°C
Flash Point 110°C (230°F)
Refractive Index 1.467
Quantity 25 g
Beilstein 1722277
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 194.32
Percent Purity 97%
Assay (Z)-isomer (nerylacetone) ≈45%
Chemical Name or Material Geranylacetone, (E)+(Z)