Läs mer
Geranylgeraniol (Natural) ~85%, TRC
Produktkod. 30240664
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30240664 100 mg
1 options
Produktkod. 30240664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG367499100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Geranylgeraniol (mixture of isomers)
CAS 24034-73-9
Molekylformel C20 H34 O
Kvantitet 100 mg
Synonym (2E,6E,10E)-3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)- 3,7,11,15-Tetramethyl-2,6,10,14-hexadecatetraen-1-ol, (E,E,E)-Geranylgeraniol, All-trans-Geranylgeraniol, trans,trans,trans-Geranylgeraniol, trans-Geranylgeraniol
InChI formel InChI=1S/C20H34O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h9,11,13,15,21H,6-8,10,12,14,16H2,1-5H3/b18-11+,19-13+,20-15+
LEDER CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CO)\C)\C)\C)C
IUPAC-namn (2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-ol
Molekylvikt (g/mol) 290.48
Formel vikt 290.261