Learn More
Germanium(IV) Oxide, Sigma-Aldrich™
Product Code. 15877890
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877890 25 g 25g
15867890 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15877890 Supplier Sigma Aldrich Supplier No. 1121770025
 Restricted Item
Beräknad leverans: 18-11-2026
Chemical Identifiers
CAS 1310-53-8
Specifications
Melting Point 1115°C
Quantity 25 g
Density 4.23 g/cm3 at 25°C
MDL Number MFCD00011030
Synonym Germanium(IV) oxide, Germanium(IV) oxide
Molecular Weight (g/mol) 104.64
Grade Research
Chemical Name or Material Germanium(IV) Oxide