missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFOD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16340-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Promotes matrix assembly.Specifications
GFOD2 | |
Polyclonal | |
Unconjugated | |
GFOD2 | |
GFOD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307801, 70575, 81577 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3B7J2, Q9CYH5 | |
Gfod2 | |
GFOD2 Fusion Protein Ag9518 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |