Beskrivning
GFR alpha-3/GDNF R alpha-3 Polyclonal specifically detects GFR alpha-3/GDNF R alpha-3 in Human samples. It is validated for ELISA.
Specifikationer
Specifikationer
| Värd art | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human GFR alpha-3/GDNF R alpha-3 (Accession#: NP_001487.2; Met 1-Trp 382) |
| Produkttyp | Antibody |
| Produktlinje | Novus |