Läs mer
GGTI298, MedChemExpress
Produktkod. 16480709
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16480709 1 mg
16435937 5 mg
16423208 10 mg
16419756 50 mg
16412676 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16480709 Leverantör Medchem Express Leverantörsnummer HY100876
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
MedChemExpress GGTI298 is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, strongly inhibiting the processing of geranylgeranylated Rap1A with little effect on processing of farnesylated Ha-Ras, with IC50 values of 3 and > 20 μM in vivo, respectively.
GGTI298 is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, strongly inhibiting the processing of geranylgeranylated Rap1A with little effect on processing of farnesylated Ha-Ras, with IC50 values of 3 and > 20 μM in vivo, respectively.
Specifikationer
Kemiskt namn eller material GGTI298
CAS 180977-44-0
Färg Gray
Anteckningar om renhetsgrad Research
Molekylformel C27H33N3O3S
Kvantitet 1 mg
Löslighetsinformation DMSO : 100 mg/mL (208.49 mM; Need ultrasonic)
LEDER CC(C)C[C@@H](C(OC)=O)NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=C3C=CC=CC3=CC=C2)=O
Molekylvikt (g/mol) 479.63
Formel vikt 479.63
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease