missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GHITM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GHITM |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825894
|
Proteintech
16296-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815894
|
Proteintech
16296-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.Specifications
GHITM | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H3K2 | |
27069 | |
GHITM Fusion Protein Ag9482 | |
Primary | |
-20°C | |
GHITM |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DERP2, GHITM, HSPC282, MICS1, My021, PTD010, TMBIM5, UNQ244/PRO281 | |
GHITM | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |