missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GHR Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GHR |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850285
|
Proteintech
20713-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840285
|
Proteintech
20713-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
may play a role in chondrocyte differentiation and positive regulation of longitudinal bond growth and.Specifications
GHR | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Porcine, Human | |
P10912, P19756 | |
2690, 397488 | |
Peptide | |
Primary | |
-20°C | |
GHR |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GH binding protein, GH receptor, GHBP, GHR, growth hormone receptor, Serum binding protein, Somatotropin receptor | |
GHR | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |