missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ GHRH Polyclonal Antibody

Beskrivning
Antibody detects endogenous levels of total GHRH.
The activated kinase acts on a variety of targets. Likely to be the GTPase effector that links the Rho-related GTPases to the JNK MAP kinase pathway. Activated by CDC42 and RAC1. Involved in dissolution of stress fibers and reorganization of focal complexes. Involved in regulation of microtubule biogenesis through phosphorylation of TBCB. Activity is inhibited in cells undergoing apoptosis, potentially due to binding of CDC2L1 and CDC2L2.
Specifikationer
Specifikationer
| Antigen | GHRH |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | Ghrh |
| Genaccessionsnr. | P01286, P09916, P16043 |
| Gene Alias | GHRF; Ghrh; GRF; growth hormone releasing hormone; growth hormone-releasing factor; Growth hormone-releasing hormone; INN; MGC119781; OTTHUMP00000030915; Sermorelin; somatocrinin; Somatoliberin; somatorelin |
| Gensymboler | Ghrh |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?