missing translation for 'onlineSavingsMsg'
Learn More

Gibberellenic Acid, TRC

Product Code. 30405969
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30405969 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405969 Supplier Toronto Research Chemicals Supplier No. TRCG377300250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Gibberellenic Acid
CAS 546-09-8
Molekylformel C19H22O6
Kvantitet 250 mg
Synonym (1α,2β,10β)-2,7-Dihydroxy-1-methyl-8-methylene-gibba-3,4a-diene-1,10-dicarboxylic Acid, 2β,7-Dihydroxy-1-methyl-8-methylene-gibba-3,4a-diene-1α,10β-dicarboxylic Acid
InChI formel InChI=1S/C19H22O6/c1-9-7-18-8-19(9,25)6-5-11(18)10-3-4-12(20)17(2,16(23)24)13(10)14(18)15(21)22/h3-4,12-14,20,25H,1,5-8H2,2H3,(H,21,22)(H,23,24)/t12-,13+,14+,17+,18-,19-/m0/s1
LEDER OCC(=O)[C@]1(OC(C)=O)[C@]2(C)[C@@H](CC1)[C@H]3[C@H]([C@@H](O)C2)[C@]4(C)C(=CC(=O)CC4)CC3
Molekylvikt (g/mol) 346.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.