Läs mer
Gibberellic Acid Acetate, TRC
Produktkod. 30397908
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30397908 100 mg
1 options
Produktkod. 30397908 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG377530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Gibberellic Acid Acetate
CAS 7648-02-4
Molekylformel 2 C11 H14 N2 . H Cl
Kvantitet 100 mg
Synonym 2-O-Acetylgibberellic acid, (1alpha,2beta,4aalpha,4bbeta,10beta)-2-(Acetyloxy)-4a,7-dihydroxy-1-methyl-8-methylene-gibb-3-ene-1,10-dicarboxylic Acid 1,4a-Lactone, 2-acetate 2beta,4a,7-Trihydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibb-3-ene-1alpha 10beta-Dicarboxylic acid 1,4a-Lactone
InChI formel InChI=1S/2C11H14N2.ClH/c2*1-8-9(6-7-12)10-4-2-3-5-11(10)13-8;/h2*2-5,13H,6-7,12H2,1H3;1H
LEDER Cl.Cc1[nH]c2ccccc2c1CCN.Cc3[nH]c4ccccc4c3CCN
IUPAC-namn 2-(2-methyl-1H-indol-3-yl)ethanamine;hydrochloride
Molekylvikt (g/mol) 384.945
Formel vikt 384.208