Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Kemiskt namn eller material | Gibberellic acid |
| CAS | 77-06-5 |
| CAS Max % | ≤ 100% |
| Beilstein | 54346 |
| Färg | Colorless |
| Smältpunkt | 233°C to 235°C (Decomposition at melting point) |
| Synonym | (3S,3aS,4S,4aS,7S,9aR,9bR,12S)-7,12-dihydroxy-3-methyl-6-methylene-2-oxoperhydro-4a,7-methano-9b,3-propenoazuleno[1,2-b]furan-4-carboxylic acid |
| Molekylvikt (g/mol) | 346.37 g/mol |
| Kvantitet | 100 mg |