Läs mer
Gibberellic Acid Methyl Ester Acetate, TRC
Produktkod. 30408046
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30408046 100 mg
1 options
Produktkod. 30408046 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG377550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Gibberellic Acid Methyl Ester Acetate
CAS 55732-63-3
Molekylformel C22H26O7
Kvantitet 100 mg
Synonym 2-(Acetyloxy)-4a,7-dihydroxy-1-methyl-8-methylene-, 1,4a-lactone, 10-methyl Ester, (1alpha,2beta,4aalpha,4bbeta,10beta)-gibb-3-ene-1,10-dicarboxylic Acid
InChI formel InChI=1S/C22H26O7/c1-11-9-20-10-21(11,26)7-5-13(20)22-8-6-14(28-12(2)23)19(3,18(25)29-22)16(22)15(20)17(24)27-4/h6,8,13-16,26H,1,5,7,9-10H2,2-4H3/t13-,14+,15-,16-,19-,20+,21+,22-/m1/s1
LEDER COC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](OC(=O)C)C=C[C@@]2(OC3=O)[C@@H]4CC[C@]5(O)C[C@]14CC5=C
Molekylvikt (g/mol) 402.44
Formel vikt 402.1679