Läs mer
Gibberellin A4, TRC
Produktkod. 30011076
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011076 10 mg
30009720 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG37745510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 468-44-0
Molekylformel C19 H24 O5
Molekylvikt (g/mol) 332.39
Synonym (1α,2β,4aα,4bβ,10β)-2,4a-Dihydroxy-1-methyl-8-methylenegibbane-1,10-dicarboxylic acid 1,4a-lactone, 4aα,4bβ-Gibbane-1α,10β-dicarboxylic acid, 2β,4a-dihydroxy-1-methyl-8-methylene-, 1,4a-lactone (8CI), Gibberellin A4 (7CI), 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibbane-1,10-dicarboxylic acid deriv., GA4, Gibberellin GA4, Gibberellin A4
LEDER C[C@]12[C@@H](O)CCC3(OC1=O)[C@@H]4CC[C@@H]5CC4(CC5=C)[C@H]([C@H]23)C(=O)O
Specifikationer
Kemiskt namn eller material Gibberellin A4
Kvantitet 10 mg
InChI formel InChI=1S/C19H24O5/c1-9-7-18-8-10(9)3-4-11(18)19-6-5-12(20)17(2,16(23)24-19)14(19)13(18)15(21)22/h10-14,20H,1,3-8H2,2H3,(H,21,22)/t10-,11-,12+,13-,14-,17-,18?,19?/m1/s1
Formel vikt 332.1624