Läs mer
Gibberellin A5, TRC
Produktkod. 30138435
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138435 100 mg
30011697 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG377465100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 561-56-8
Molekylformel C19 H22 O5
Molekylvikt (g/mol) 330.38
Synonym 4a,7-Dihydroxy-1-methyl-8-methylene-4aα,4bβ-Gibb-2-ene-1α,10β-dicarboxylic Acid 1,4a-Lactone, GA5, GA5 (Gibberellin)
LEDER C[C@]12C=CC[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O
Specifikationer
Kemiskt namn eller material Gibberellin A5
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C19H22O5/c1-10-8-17-9-18(10,23)7-4-11(17)19-6-3-5-16(2,15(22)24-19)13(19)12(17)14(20)21/h3,5,11-13,23H,1,4,6-9H2,2H3,(H,20,21)/t11-,12-,13-,16-,17+,18+,19-/m1/s1
Formel vikt 330.1467
Procent renhet >95%