missing translation for 'onlineSavingsMsg'
Learn More

Gibberellin A9 Methyl Ester, TRC

Product Code. 30240729
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30240729 50 mg
30240596 100 mg
30240662 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.
Product Code. 30240729 Supplier Toronto Research Chemicals Supplier No. TRCG37732550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains plant protection products according to Regulation (EC) No 1107/2009, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2112-08-5
Molekylformel C20H26O4
Molekylvikt (g/mol) 330.42
Synonym GA9 Methyl Ester, 1,4a-Lactone-4a-hydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibbane-1alpha,10beta-dicarboxylic Acid Methyl Ester;
LEDER COC(=O)[C@H]1[C@@H]2[C@@]3(C)CCC[C@@]2(OC3=O)[C@@H]4CC[C@@H]5C[C@]14CC5=C

Specifications

Kemiskt namn eller material Gibberellin A9 Methyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C20H26O4/c1-11-9-19-10-12(11)5-6-13(19)20-8-4-7-18(2,17(22)24-20)15(20)14(19)16(21)23-3/h12-15H,1,4-10H2,2-3H3/t12-,13-,14-,15-,18-,19+,20-/m1/s1
Formel vikt 330.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.