Läs mer
Gibberellin A9, TRC
Produktkod. 17967824
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17967824 1 mg
17908494 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17967824 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG3775251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 427-77-0
Molekylformel C19H24O4
Molekylvikt (g/mol) 316.39
Synonym Dodecahydro-4a-hydroxy-1-methyl-8-methylene-1H-7,9a-methanobenz[a]azulene-1,10-dicarboxylic Acid 1,4a-Lactone, 4a-Hydroxy-1-methyl-8-methylene-4aalpha,4bbeta-gibbane-1alpha,10beta-dicarboxylic Acid 1,4a-Lactone, Gibberellin A9, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene, gibbane-1,10-dicarboxylic Acid Deriv., GA9
LEDER C[C@]12CCC[C@@]3(OC1=O)[C@@H]4CC[C@@H]5C[C@]4(CC5=C)[C@H]([C@H]23)C(=O)O
Specifikationer
Kemiskt namn eller material Gibberellin A9
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C19H24O4/c1-10-8-18-9-11(10)4-5-12(18)19-7-3-6-17(2,16(22)23-19)14(19)13(18)15(20)21/h11-14H,1,3-9H2,2H3,(H,20,21)/t11-,12-,13-,14-,17-,18+,19-/m1/s1
Formel vikt 316.1675
Procent renhet >95%