missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Cystatin C Recombinant Human Protein
Recombinant Human Cyststatin-C is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHP0044
Additional Details : Weight : 0.00400kg
Description
Cystatin-C is a cysteine proteinase inhibitor that is involved in kidney function, cardiovascular disease and neurologic disorders. Cystatin-C has been shown to colocalize with amyloid beta in Alzheimer's disease patients.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by measuring the inhibition of papin’s protease activity
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
10 μg | |
Human | |
Store in freezer (-5 to -30°C). | |
1.5 μM | |
< 1 EU/ml | |
Dry Ice | |
Proteases Peptidases & Inhibitors | |
Lyophilized | |
Oncology, Gene Regulation, Virology, Cardiovascular Research |
13 kDa | |
Cystatin C | |
Recombinant | |
Carrier-Free | |
Colorimetric assay using L-BAPA as substrate | |
10 μg | |
Mammalian | |
Recombinant, Ligand | |
Proteinase Inhibitor | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.