Learn More
Gibco™ Human IFN-alpha 2 (alpha 2b) (Carrier-Free) Recombinant Protein
Human IFN-alpha 2 (alpha 2b) (Carrier-Free) Recombinant Protein
Brand: Gibco™ PHC4064
Additional Details : Weight : 0.02000kg
Description
Human IFN-alpha 2 (alpha 2b) (Carrier-Free) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IFN-α is involved in anti-viral and anti-parasitic activitites.Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
19.2 kDa | |
IFN-α2b | |
Store in ultra-cold freezer (-68 to -85°C). | |
Dry Ice | |
Cytokines & Receptors | |
Liquid | |
Immunology, Virology, Oncology, Signal Transduction | |
95 % |
3440 | |
1 x 10 exp. 5 units | |
Human | |
10^5 units | |
E. coli | |
Recombinant, Ligand | |
Interferons | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.