Learn More
Gibco™ Human IFN-alpha A (alpha 2a) Recombinant Protein
Human IFN-alpha A (alpha 2a) Recombinant Protein
Brand: Gibco™ PHC4014
Additional Details : Weight : 0.50000kg
Description
Human IFN-alpha A (alpha 2a) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IFN-α is involved in anti-viral and anti-parasitic activitites.Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
19.2 kDa | |
5 ug | |
Human | |
Carrier-Free | |
Dry Ice | |
Cytokines & Receptors | |
Liquid | |
Immunology, Virology, Oncology, Signal Transduction | |
95 % |
3438 | |
Sequential Chromatography | |
IFN-α | |
Store in ultra-cold freezer (-68 to -85°C). | |
5 μg | |
E. coli | |
Recombinant, Ligand | |
Interferons | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.