missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL17 Recombinant Mouse Protein

Recombinant IL17 is a bioactive protein intended for use in cell culture applications

2205.00 SEK - 3185.00 SEK

Specifications

Purity 95 %
Purification Method Sequential Chromatography
Gene Alias IL-17
Gene ID (Entrez) 16171
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10029472
View Documents
Gibco™
PMC0174
10μg
3185.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment
11-11-2020
Add to basket
 
10242533
View Documents
Gibco™
PMC0175
25μg
2205.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment
11-11-2020
Add to basket
 
Description

Description

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice.

Specifications

Specifications

95 %
Sequential Chromatography
IL-17
16171
SDS

For Research Use Only. Not for use in diagnostic procedures.