Learn More
Gibco™ IL6 Recombinant Mouse Protein
Recombinant Mouse Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications
2220.00 SEK - 4340.00 SEK
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 16193 |
Molecular Weight (g/mol) | ∼20 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10262353
|
Gibco™
PMC0065 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10689963
|
Gibco™
PMC0066 |
25μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse B9 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
∼20 kDa | |
IL-6 | |
Recombinant | |
Carrier-Free | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
16193 | |
Sequential Chromatography | |
Mouse | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Inflammation, Bone Research | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.