missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ LEFTY-A Recombinant Human Protein
Recombinant Human LEFTY-A is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC1714
Additional Details : Weight : 0.00400kg
Description
LEFTY-A are TGF-β ligands that inhibit TGF-β signalling. LEFTY-A has been shown to induce mitogen activated protein kinase activity.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose dependent ability to inhibit BMP4 induction of alkaline phosphatase production in the MC-3T3-E1 cell line
- Proven compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
10 μg | |
Human | |
Store in freezer (-5 to -30°C). | |
40 ng/ml | |
< 1 EU/ml | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Oncology |
38 kDa | |
LEFTY-A | |
Recombinant | |
Carrier-Free | |
Inhibition of BMP-4 induction of alkaline phosphatase production in MC-3T3-E1 cell line | |
10 μg | |
Mammalian | |
Recombinant, Ligand | |
TGFs (Transforming Growth Factors) | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.