missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL18 (MIP-4) Recombinant Protein
Human CCL18 (MIP-4) Recombinant Protein
Brand: Gibco™ PHC1254
Additional Details : Weight : 0.50000kg
Description
Human CCL18 (MIP-4) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
MIP-4 (Macrophage inflammatory protein-4) is a chemokine and is also known as CCL18 and PARC.Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
7.8 kDa | |
10 ug | |
69 aa | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Chemokines & Receptors | |
Lyophilized | |
Immunology, Inflammation, Signal Transduction, Neurobiology | |
98 % |
6362 | |
Sequential Chromatography | |
MIP-4 | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
MIP (Macrophage Inflammatory Protein) Chemokines | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.