Learn More
Gibco™ Human Osteoprotegerin (TNFRSF11B) Recombinant Protein
Human Osteoprotegerin (TNFRSF11B) Recombinant Protein
Brand: Gibco™ PHC1684
Additional Details : Weight : 0.50000kg
Description
Human Osteoprotegerin (TNFRSF11B) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Osteoprotegerin is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
20 kDa | |
50 ug | |
174 aa | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Bone Research, Signal Transduction | |
98 % |
4982 | |
Sequential Chromatography | |
Osteoprotegerin | |
Human | |
Carrier-Free | |
50 μg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.