Learn More
Gibco™ TPO Recombinant Mouse Protein
Recombinant TPO is a bioactive protein intended for use in cell culture applications
Brand:  Gibco™ PMC1145
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10009422
  • 4015.00 SEK / Each
Estimated Shipment date
from Supplier 05-03-2019
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Specifications
Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
22018
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
17kDa
95%
Mouse
Carrier Protein
TPO
TPO
Growth Factors & Receptors