Läs mer
GIBH-130, MedChemExpress
Produktkod. 16473618
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
2mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16473618 10 mg -
16464706 100 mg -
16450066 2 mg 2mg
16436347 5 mg 5mg
16410017 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16473618 Leverantör Medchem Express Leverantörsnummer HY101860
MedChemExpress GIBH-130 is an effective inhibitor of neuroinflammation. GIBH-130 significantly suppresses the IL-1β secretion by activated microglia (IC50=3.4 nM).
GIBH-130 is an effective inhibitor of neuroinflammation. GIBH-130 significantly suppresses the IL-1β secretion by activated microglia (IC50=3.4 nM).
Specifikationer
Kemiskt namn eller material GIBH-130
Fysisk form Solid
CAS 1252608-59-5
Färg Pink
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H20N6O
För användning med (applikation) Neuroscience-Neurodegeneration
Löslighetsinformation DMSO : 50 mg/mL (138.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER O=C(C1=NN=C(C2=CC=CC=C2)C=C1C)N3CCN(C4=NC=CC=N4)CC3
Molekylvikt (g/mol) 360.41
Formel vikt 360.41
Procent renhet 99.95%