Learn More
Macherey-Nagel™ Gilson ASPEC Chromabond™ SiOH Polypropylene SPE Column
For automated systems or vacuum manifolds
Brand: Macherey-Nagel™ 730073ASP
Additional Details : Weight : 1.00000kg
Description
- Automated systems or CHROMABOND™ vacuum manifolds
- Polypropylene column, polyethylene filter element
- High loading capacity and separation efficiency
- Constant back pressure, airtight sealing
- Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
3 mL | |
500 mg | |
Cap |
SPE | |
Solid Phase Extraction Column |