Learn More
Macherey-Nagel™ Gilson ASPEC Chromabond™ SiOH Polypropylene SPE Column
For automated systems or vacuum manifolds
Brand:  Macherey-Nagel™ 730075ASP
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12874451
  • 5825.00 SEK / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Automated systems or CHROMABOND™ vacuum manifolds
  • Polypropylene column, polyethylene filter element
  • High loading capacity and separation efficiency
  • Constant back pressure, airtight sealing
  • Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
Specifications
6 mL
1 g
Cap
SPE
Solid Phase Extraction Column