Läs mer
Invitrogen™ Gilvetmab Recombinant Canine Monoclonal Antibody
Canine Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542149
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Gilvetmab is a biosimilar that targets PDCD1.
Specifikationer
| Gilvetmab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AH-PD1 | |
| Canine (Canis lupus familiaris) PDCD1/PD1/CD279. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 3.4 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Canine | |
| Protein A/G | |
| RUO | |
| Canine | |
| Antibody | |
| IgG2 κ |